missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSLP Monoclonal Antibody (K49024_1G12), Invitrogen™

Numéro de catalogue. MA556148
Modifier l'affichage
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA556148 200 μL
MA555768 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA556148 Fournisseur Invitrogen™ Code fournisseur MA556148
Il en reste null

Mouse Monoclonal Antibody

Sequence of this protein is as follows: YDFTNCDFEK IKAAYLSTIS KDLITYMSGT KSTEFNNTVS CSNRPHCLTE IQSLTFNPTA GCASLAKEMF AMKTKAALAI WCPGYSETQI NATQAMKKRR KRKVTTNKCL EQVSQLQGLW RRFNRPLLKQ Q

TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.

Spécifications

Antigen TSLP
Applications Western Blot
Classification Monoclonal
Clone K49024_1G12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene TSLP
Gene Accession No. Q969D9
Gene Alias Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP
Gene Symbols TSLP
Host Species Mouse
Immunogen Recombinant human TSLP (immunogen range is 29-159).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85480
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Afficher plus de résultats Afficher moins de résultats