missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSKgel™ Ether-5PW HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™
An excellent choice for separating hydrophobic molecules such as membrane proteins, or monoclonal antibodies such as IgG or IgM.
Supplier: Merck Emd Millipore 814014
Description
- Among the TSKgel HIC Columns, TSKgel Ether-5PW has intermediate hydrophobity
- An excellent choice for separating hydrophobic molecules such as membrane proteins or monoclonal antibodies such as IgG or IgM
- Features a 1000 Å pore size in a polymethacrylate base material
- Column is stable in either acid or caustic cleaning regimens
- Provides excellent access to larger molecules with low diffusion coefficients
- Both hydrophobic interaction chromatography (HIC) and reversed-phase liquid chromatography separate on the basis of protein hydrophobicity allowing for selective binding, and desorption of proteins
- Recommended for purifying more hydrophobic proteins
- Salt Concentration: ≤4 M
Specifications
| 8 mm | |
| 7.5 cm | |
| 10 μm | |
| ≤ 50% Organic Solvent | |
| Ether | |
| Ether Phase/Hydroxylated Methacrylic Polymer | |
| HIC HPLC |
| 1.2 mL/min | |
| 20 bar | |
| 2 to 12 | |
| 10°C to 50°C | |
| TSKgel | |
| Glass |
TSKgel is a registered trademark of Tosoh Corporation