missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ Ether-5PW HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. p-7089815
Change view
Click to view available options
Diameter (Metric):
2 mm
5 mm
7.5 mm
8 mm
Material:
Glass
Stainless Steel
Stainless Steel Hardware
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Material
111021844 5 mm Glass
111021845 8 mm Glass
111021973 2 mm Stainless Steel Hardware
111021814 7.5 mm Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111021844 Supplier Merck Emd Millipore Supplier No. 814013
Only null left

An excellent choice for separating hydrophobic molecules such as membrane proteins, or monoclonal antibodies such as IgG or IgM.

  • Among the TSKgel HIC Columns, TSKgel Ether-5PW has intermediate hydrophobity
  • An excellent choice for separating hydrophobic molecules such as membrane proteins or monoclonal antibodies such as IgG or IgM
  • Features a 1000 Å pore size in a polymethacrylate base material
  • Column is stable in either acid or caustic cleaning regimens
  • Provides excellent access to larger molecules with low diffusion coefficients
  • Both hydrophobic interaction chromatography (HIC) and reversed-phase liquid chromatography separate on the basis of protein hydrophobicity allowing for selective binding, and desorption of proteins
  • Recommended for purifying more hydrophobic proteins
  • Salt Concentration: ≤4 M

Specifications

Diameter (Metric) 5 mm
Flow Rate 1 mL/min
Length (Metric) 5 cm
Max. Pressure 20 bar
Particle Size 10 μm
pH 2 to 12
Solvents ≤ 50% Organic Solvent
Temperature 10°C to 50°C
Stationary Phase Ether
Product Line TSKgel
Matrix Ether Phase/Hydroxylated Methacrylic Polymer
Material Glass
Technique HIC HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation