Learn More
New England Biolab Ltd Template Switching RT Enzyme Mix 20 reactions
Numéro de catalogue. NC1732525
Encompass
The reverse transcriptase (RT) in the Template Switching RT Enzyme Mix adds a few non-templated nucleotides after it reaches the 5' end of the RNA template. These non-templated nucleotides can anneal to a template switching oligo (TSO) with a known sequence, prompting the reverse transcriptase to switch template from RNA to the TSO. The resulting cDNA contains a known sequence (complementary to the sequence of the TSO) attached to the 3' end. This feature can be utilized in a variety of downstream applications, such as cDNA amplification, 5' RACE (rapid amplification of cDNA ends), and 2nd strand cDNA synthesis.
Numéro de catalogue. NC1732525 Fournisseur New England Biolab Ltd Code fournisseur M0466S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The reverse transcriptase (RT) in the Template Switching RT Enzyme Mix adds a few non-templated nucleotides after it reaches the 5' end of the RNA template. These non-templated nucleotides can anneal to a template switching oligo (TSO) with a known sequence, prompting the reverse transcriptase to switch template from RNA to the TSO. The resulting cDNA contains a known sequence (complementary to the sequence of the TSO) attached to the 3' end. This feature can be utilized in a variety of downstream applications, such as cDNA amplification, 5' RACE (rapid amplification of cDNA ends), and 2nd strand cDNA synthesis.