Learn More
Worthington Biochemical Trypsin, TPCK Treated, 100mg
Numéro de catalogue. NC9783694
Encompass
A chromatographically purified, diafiltered, lyophilized powder that has been treated with L-(tosylamido-2-phenyl) ethyl chloromethyl ketone (TPCK) to inhibit contaminating chymotryptic activity [Kostka, V., and Carpenter, F.: JBC, 239, 1799 (1964)]. Store at 2-8C. Protect from moisture.
Numéro de catalogue. NC9783694 Fournisseur Worthington Biochemical Code fournisseur LS003740
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
A chromatographically purified, diafiltered, lyophilized powder that has been treated with L-(tosylamido-2-phenyl) ethyl chloromethyl ketone (TPCK) to inhibit contaminating chymotryptic activity [Kostka, V., and Carpenter, F.: JBC, 239, 1799 (1964)]. Store at 2-8C. Protect from moisture.