Learn More
StatLab™ TruCut™ Microtome Blades
Description
- Blade edge is slowly refined, sharpened and optimized to maximize hardness, sharpness and durability
- Made in Japan with a proprietary surface ionization process
Specifications
Specifications
| Profile | High |
| Disposable | Single-use |
| Quantity | 50 Pk. |