Learn More
Edvotek™ TruBlu™ 2 Blue/White Transilluminator
Catalog No. S42878
Change view
Click to view available options
Wavelength:
470 nm
Catalog No. Wavelength
S42878 470 nm
1 options
This item is intended for educational purposes only.
Catalog No. S42878 Supplier Edvotek™ Supplier No. 557
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This item is intended for educational purposes only.
For viewing DNA gels stained with SYBR™ Safe eliminating the need for UV light or ethidium bromide. Edvotek TruBlu 2 Blue Light Transilluminator is optimized to fit Edvotek Gels and the high intensity control and orange lid ensure superior visualization.
  • Developed in concert with the inventor of the technology under license from Clare Chemical Research, Inc
  • Viewing Area: 27 x 15 cm
  • Optimized for Edvotek Gels
  • Blue Light High Intensity Control
  • Orange Contrast Lid
  • Vivid results
  • US Patent Nos. 6,198,107, 6,512,236, 6,914,250
  • EP Patent No. 0 965 034
  • Made in USA
Specifications
Product Type Transilluminator
For Use With (Application) Viewing DNA gels stained with SYBR Safe
Wavelength 470 nm
Lighting Type LED (Blue and White)