missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troxerutin 90.0+%, TCI America™
$102.52 - $307.57
Chemical Identifiers
| CAS | 7085-55-4 |
|---|---|
| Molecular Formula | C33H42O19 |
| Molecular Weight (g/mol) | 742.68 |
| MDL Number | MFCD00893813 |
| InChI Key | IYVFNTXFRYQLRP-VVSTWUKXSA-N |
| Synonym | 3′,4′,7-Tris[O-(2-hydroxyethyl)]rutin, Trihydroxyethylrutin |
| PubChem CID | 5486699 |
| IUPAC Name | 2-[3,4-bis(2-hydroxyethoxy)phenyl]-5-hydroxy-7-(2-hydroxyethoxy)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
| SMILES | CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=C(OC4=CC(=CC(=C4C3=O)O)OCCO)C5=CC(=C(C=C5)OCCO)OCCO)O)O)O)O)O)O |
Chemical Identifiers
| 7085-55-4 | |
| 742.68 | |
| IYVFNTXFRYQLRP-VVSTWUKXSA-N | |
| 5486699 | |
| CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=C(OC4=CC(=CC(=C4C3=O)O)OCCO)C5=CC(=C(C=C5)OCCO)OCCO)O)O)O)O)O)O |
| C33H42O19 | |
| MFCD00893813 | |
| 3′,4′,7-Tris[O-(2-hydroxyethyl)]rutin, Trihydroxyethylrutin | |
| 2-[3,4-bis(2-hydroxyethoxy)phenyl]-5-hydroxy-7-(2-hydroxyethoxy)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
Specifications
| 7085-55-4 | |
| C33H42O19 | |
| 5 g | |
| IYVFNTXFRYQLRP-VVSTWUKXSA-N | |
| 2-[3,4-bis(2-hydroxyethoxy)phenyl]-5-hydroxy-7-(2-hydroxyethoxy)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one | |
| 5486699 | |
| ≥90.0% (HPLC) | |
| Troxerutin |
| 186°C | |
| MFCD00893813 | |
| 3′,4′,7-Tris[O-(2-hydroxyethyl)]rutin, Trihydroxyethylrutin | |
| CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=C(OC4=CC(=CC(=C4C3=O)O)OCCO)C5=CC(=C(C=C5)OCCO)OCCO)O)O)O)O)O)O | |
| 742.68 | |
| 742.68 | |
| Powder |
Safety and Handling
RTECSNumber : LK8331500
TSCA : No
Recommended Storage : Freezer