missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Troxerutin | |
| 186°C | |
| MFCD00893813 | |
| 3′,4′,7-Tris[O-(2-hydroxyethyl)]rutin, Trihydroxyethylrutin | |
| CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=C(OC4=CC(=CC(=C4C3=O)O)OCCO)C5=CC(=C(C=C5)OCCO)OCCO)O)O)O)O)O)O | |
| 742.68 | |
| 742.68 | |
| Powder |
| 7085-55-4 | |
| C33H42O19 | |
| 25 g | |
| IYVFNTXFRYQLRP-VVSTWUKXSA-N | |
| 2-[3,4-bis(2-hydroxyethoxy)phenyl]-5-hydroxy-7-(2-hydroxyethoxy)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one | |
| 5486699 | |
| ≥90.0% (HPLC) |
Chemical Identifiers
| 7085-55-4 | |
| 742.68 | |
| IYVFNTXFRYQLRP-VVSTWUKXSA-N | |
| 5486699 | |
| CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=C(OC4=CC(=CC(=C4C3=O)O)OCCO)C5=CC(=C(C=C5)OCCO)OCCO)O)O)O)O)O)O |
| C33H42O19 | |
| MFCD00893813 | |
| 3′,4′,7-Tris[O-(2-hydroxyethyl)]rutin, Trihydroxyethylrutin | |
| 2-[3,4-bis(2-hydroxyethoxy)phenyl]-5-hydroxy-7-(2-hydroxyethoxy)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
Safety and Handling
RTECSNumber : LK8331500
TSCA : No
Recommended Storage : Freezer