missing translation for 'onlineSavingsMsg'
Learn More

Troxerutin 90.0+%, TCI America™

Catalog No. t35415g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35415G 5 g
T354125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35415G Supplier TCI America Supplier No. T35415G

Chemical Identifiers

CAS 7085-55-4
Molecular Formula C33H42O19
Molecular Weight (g/mol) 742.68
MDL Number MFCD00893813
InChI Key IYVFNTXFRYQLRP-VVSTWUKXSA-N
Synonym 3′,4′,7-Tris[O-(2-hydroxyethyl)]rutin, Trihydroxyethylrutin
PubChem CID 5486699
IUPAC Name 2-[3,4-bis(2-hydroxyethoxy)phenyl]-5-hydroxy-7-(2-hydroxyethoxy)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
SMILES CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=C(OC4=CC(=CC(=C4C3=O)O)OCCO)C5=CC(=C(C=C5)OCCO)OCCO)O)O)O)O)O)O

Specifications

Melting Point 186°C
Quantity 5 g
Formula Weight 742.68
Percent Purity ≥90.0% (HPLC)
Physical Form Powder
Chemical Name or Material Troxerutin
RTECSNumber LK8331500
TSCA No
Recommended Storage Freezer