CAS RN 7085-55-4
IUPAC Name:
2-[3,4-bis(2-hydroxyethoxy)phenyl]-5-hydroxy-7-(2-hydroxyethoxy)-3-[(3,4,5-trihydroxy-6-{[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]methyl}oxan-2-yl)oxy]-4H-chromen-4-one
Synonyms:
troxerutin
Molecular Weight (g/mol):
742.68
Molecular Formula:
C33H42O19
InChi Key:
IYVFNTXFRYQLRP-UHFFFAOYNA-N
SMILES:
CC1OC(OCC2OC(OC3=C(OC4=CC(OCCO)=CC(O)=C4C3=O)C3=CC=C(OCCO)C(OCCO)=C3)C(O)C(O)C2O)C(O)C(O)C1O
1
–
1
of
1
results
1
Troxerutin 90.0+%, TCI America™
CAS: 7085-55-4 Molecular Formula: C33H42O19 Molecular Weight (g/mol): 742.68 MDL Number: MFCD00893813 InChI Key: IYVFNTXFRYQLRP-VVSTWUKXSA-N Synonym: 3′,4′,7-Tris[O-(2-hydroxyethyl)]rutin, Trihydroxyethylrutin PubChem CID: 5486699 IUPAC Name: 2-[3,4-bis(2-hydroxyethoxy)phenyl]-5-hydroxy-7-(2-hydroxyethoxy)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one SMILES: CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=C(OC4=CC(=CC(=C4C3=O)O)OCCO)C5=CC(=C(C=C5)OCCO)OCCO)O)O)O)O)O)O