missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tropolone Tosylate 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 160°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 276.31 |
| Percent Purity | ≥99.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tropolone Tosylate |
Safety and Handling
| TSCA | No |