Learn More
Hengar Boiling Granules
Catalog No. 02215513
Change view
Click to view available options
Type:
Boiling Granules, No. 12
Plain Boiling Granules
Plain Micro Granules
Selenized Boiling Granules
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
02215513 Plain Boiling Granules
HEN132C Selenized Boiling Granules
HEN133B Boiling Granules, No. 12
02215521 Plain Micro Granules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02215513 Supplier Hengar Supplier No. 901800
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Henger Granules effectively prevent superheating with its attendant dangerous explosive vaporization commonly known as bumping
  • Granules allow for a steady, copious stream of bubbles to escape without spasmodic splatter
  • Evaporation or distillation proceeds quietly and smoothly
  • Cannot contaminate the liquid
  • Reduced flask breakage
  • Minimized watching time
  • Four mesh granule size
  • Quantity: 500g
Specifications
Type Plain Boiling Granules
Material Amphoteric Alundum
Description No. 4 Mesh
Quantity 500 g