missing translation for 'onlineSavingsMsg'
Learn More

Trizma™ base, Certified by BAM, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111015522
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015522 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015522 Supplier Merck Emd Millipore Supplier No. 9344050G

Specifications

Product Line Trizma
CAS 77-86-1
Physical Form Neat
Linear Formula NH2 C(CH2 OH)3
MDL Number MFCD00004679
pH 10.5 to 12
Melting Point 167°C to 172°C
Boiling Point 219°C to 220°C (10 mmHg)
Synonym 2-Amino-2-(hydroxymethyl)-1,3-propanediol; THAM; Tris base; Tris(hydroxymethyl)aminomethane; Trometamol
Solubility Soluble in water
Type Certified Reference Material
Quantity 50 g
Formula Weight 121.14
Percent Purity ≥99.5%
Grade Certified Reference Material
Show More Show Less

Trizma is a registered trademark of Sigma-Aldrich Co. LLC