missing translation for 'onlineSavingsMsg'
Learn More

Trityl Chloride 98.0+%, TCI America™

Catalog No. C0308500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0308500G 500 g
C030825G 25 g
C0308100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0308500G Supplier TCI America Supplier No. C0308500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76-83-5
Molecular Formula C19H15Cl
Molecular Weight (g/mol) 278.78
MDL Number MFCD00000813,MFCD00284810
InChI Key JBWKIWSBJXDJDT-UHFFFAOYSA-N
Synonym trityl chloride, triphenylmethyl chloride, triphenylchloromethane, chlorotriphenylmethane, chloromethanetriyl tribenzene, triphenylmethylchloride, methane, chlorotriphenyl, triphenyl chloromethane, chlorodiphenylmethyl benzene, benzene, 1,1',1-chloromethylidyne tris
PubChem CID 6456
IUPAC Name (chlorodiphenylmethyl)benzene
SMILES ClC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 113°C
Color White-Yellow
Boiling Point 310°C
Quantity 500 g
UN Number 3261
Formula Weight 278.78
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trityl Chloride
EINECSNumber (9)-0897
RTECSNumber PA6450000
TSCA Yes
Recommended Storage Refrigerator