missing translation for 'onlineSavingsMsg'
Learn More

Triton™ X-114, MP Biomedicals™

Catalog No. ICN19397190
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19397190 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19397190 Supplier MP Biomedicals Inc Supplier No. 0219397190

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Triton™ X-114 is a surfactant used in industrial and household detergent applications and in emulsifying agents.

  • Triton™ X-114 is used in biochemical applications to solubilize and separate proteins due to its low cloud point (23°C).
  • At temperatures above the cloud point, detergent solutions separate into aqueous and detergent-enriched phases.
  • This makes Triton™ X-114 particularly useful in separating lipophilic proteins from hydrophilic proteins.

Chemical Identifiers

CAS 9036-19-5

Specifications

Color Light Yellow
Physical Form Liquid
Density 8.8 lb./gal
Quantity 500 mL
Packaging Plastic Container
Specific Gravity 1.054 g/ml
Molecular Formula (C2H4O)x · C14H22O (x= 7-8)
Molecular Weight (g/mol) 294.435 g/mol
Chemical Name or Material Triton™ X-114
Recommended Storage 15 to 30°C

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.