missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Triton™ X-114

Catalog No. AC422365000
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422365000 500 mL Glass bottle
AC422360025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC422365000 Supplier Thermo Scientific Chemicals Supplier No. 422365000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9002-93-1 | C16H26O2 | 250.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Polyoxyethylene(8) octylphenyl ether
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Undesignated
Physical Form Liquid
Boiling Point >200.0°C
Cloud Point 22° to 28°C (1% in aq. soln.)
Density 1.0550g/mL
pH 6 (5% aq. soln.)
Quantity 500 mL
Flash Point 254°C
Infrared Spectrum Authentic
Packaging Glass bottle
Pour Point pour point: -9°C
Refractive Index 1.4880 to 1.4950
Specific Gravity 1.055
Viscosity 260 mPa.s (25°C)
Merck Index 15, 6850
Solubility Information Solubility in water: soluble. Other solubilities: miscible in most polar organic solvents and, aromatic hydrocarbons,, insoluble in aliphatic hydrocarbons.
Formula Weight 537
Chemical Name or Material Triton™ X-114
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Causes serious eye damage.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber MD0907600
TSCA TSCA

RUO – Research Use Only