missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Triton™ X-114

CAS: 9002-93-1 | C16H26O2 | 250.38 g/mol

Supplier:  Thermo Scientific Chemicals 422360025

Quantity 2.5 L
Packaging Glass bottle

 Afficher plus de versions de ce produit

Numéro de catalogue. AC422360025


Il en reste null

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

9002-93-1
250.38
JYCQQPHGFMYQCF-UHFFFAOYSA-N
5590
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
C16H26O2
MFCD00132505
Polyoxyethylene(8) octylphenyl ether
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol

Spécifications

Triton™ X-114
9002-93-1
>200.0°C
1.0550g/mL
2.5 L
Authentic
pour point: -9°C
1.055
260 mPa.s (25°C)
15, 6850
Solubility in water: soluble. Other solubilities: miscible in most polar organic solvents and, aromatic hydrocarbons,, insoluble in aliphatic hydrocarbons.
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
250.38
537
Undesignated
Liquid
22° to 28°C (1% in aq. soln.)
6 (5% aq. soln.)
254°C
Glass bottle
1.4880 to 1.4950
C16H26O2
MFCD00132505
Polyoxyethylene(8) octylphenyl ether
JYCQQPHGFMYQCF-UHFFFAOYSA-N
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethanol
5590
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS P Statement
Causes serious eye damage.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

RTECSNumber : MD0907600

TSCA : TSCA

FDS
Documents

Documents

Promotions

Promotions