missing translation for 'onlineSavingsMsg'
Learn More

Triton X-100™, Wetting Agent, LabChem™

Catalog No. LC262801
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC262801 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC262801 Supplier LabChem Supplier No. LC262801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9036-19-5

Specifications

Color Colorless to Tan
Physical Form Liquid
Boiling Point 177°C
CAS Max % 100
Decomposition Information Carbon monoxide ; Carbon dioxide
Density 1050kg/m3
Quantity 500 mL
Flash Point 110°C
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1050kg/m3
Linear Formula C18H30O3
Solubility Information Soluble in water
Formula Weight 294.43
Grade Reagent
Chemical Name or Material Triton X-100™, Wetting Agent
Show More Show Less
Recommended Storage Room Temperature