missing translation for 'onlineSavingsMsg'
Learn More

Triton X-100, For Analysis, MilliporeSigma™

Catalog No. M1086032500
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1086032500 2.5 L Glass Bottle
M1086031000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1086032500 Supplier Technidata Supplier No. 1.08603.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9036-19-5
Molecular Formula C8H17C6H4(OCH2CH2)nOH
Synonym Octoxynol

Specifications

Color Colorless
Physical Form Liquid
Boiling Point >200°C (1013hPa)
Density 1.07 g/3 (20 °C)
pH 5.0 to 8.0 (10 g/L, H2O, 20°C)
Quantity 2.5 L
Flash Point 251°C
Packaging Glass Bottle
Chemical Name or Material Ether
RTECSNumber MD0907700