missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Triton™ X-100 Surfact-Amps™ Detergent Solution

Numéro de catalogue. pi28314
Change view
Click to view available options
Quantity:
50 mL
250 mL
6 x 10 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PI28314 6 x 10 mL
PI85111 50 mL
PI85112 250 mL
PI28313 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. PI28314 Fournisseur Thermo Scientific™ Code fournisseur LSG28314
Il en reste null

Thermo Scientific Triton X-100 Surfact-Amps Detergent Solution is a highly-purified Triton™ X-100 detergent stabilized as a 10% solution (w/v) in variable sizes packaged under nitrogen in 10 mL glass ampules or three sizes of non-leaching HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Features of Triton X-100 Surfact-Amps Detergent Solution:

  • Triton X-100—a nonionic detergent for use in various protein methods
  • Accurate—precise 10% detergent solution in ultrapure water
  • Easy-to-use—solution is simple to dispense and dilute for use
  • Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
  • Stable—packaged under inert nitrogen gas in glass ampules or HDPE bottles

Specifications for Triton X-100 Surfact-Amps Detergent Solution:

  • Visual: Clear to slightly hazy, colorless solution, free of particulate matter.
  • Concentration: 10.0±1.0%
  • Oxidants: ≤1.0μeq/mL
  • Carbonyls: ≤1.0μeq/mL
  • Suspended Solids: Residue present must not exceed Residue Reference.

Properties of Triton X-100:

  • Molecular Weight: 647 g
  • Detergent Class: Nonionic polyoxyethylene surfactant
  • Aggregation Number: 140
  • Micelle Molecular Weight: 90,000 g
  • Critical Micelle Concentration (CMC): 0.24 mM (0.0155% w/v)
  • Cloud Point: 64°C
  • Dialyzable: No

Triton X-100 is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins.

Related Products

  • Surfact-Amps™ Detergent Sampler (Cat. No. 28340)

Spécifications

Product Type Detergent
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.
Form Liquid
Product Line Surfact-Amps, Triton
Reagent Type Detergent Solution
Quantity 6 x 10 mL

For Research Use Only. Not for use in diagnostic procedures.