missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Triton X-100 Detergent, Molecular biology grade, Calbiochem™,
Specifications
Specifications
| Color | Undesignated |
| Quantity | 50 mL |
| Format | Plastic Ampoule |
| pH | 6.0 to 8.0 |