missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Triton X-100 Detergent, Calbiochem™,

Catalog No. 6484623KG
Change view
Click to view available options
Quantity:
1 kg
3 kg
Packaging:
Fiber Carton
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6484623KG 3 kg Fiber Carton
6484621KG 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6484623KG Supplier MilliporeSigma™ Supplier No. 6484623KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 9002-93-1
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
MDL Number MFCD00132505
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
Synonym Polyethylene Glycol-p-isooctylphenyl Ether
PubChem CID 5590
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1

Specifications

Color Colorless to Yellow
Quantity 3 kg
Packaging Fiber Carton