missing translation for 'onlineSavingsMsg'
Learn More

Triton(R) X-100, Spectrum™ Chemical

Catalog No. 18607597
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Drum
Poly Bottle
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607597 4 L Poly Bottle
18607598 20 L Steel Can
18607599 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18607597 Supplier Spectrum Chemical Mfg Cor Supplier No. TR1354LTPL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TR135, 9036-19-5, C14H21(C2H4O)9-10OH

Spectrum™ Chemical Triton X-100

Chemical Identifiers

CAS 9036-19-5
Molecular Formula C18H30O3
Molecular Weight (g/mol) 294.44
InChI Key LBCZOTMMGHGTPH-UHFFFAOYSA-N
IUPAC Name 2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethan-1-ol
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCOCCO)C=C1

Specifications

CAS Max % 1
Quantity 4 L
Packaging Poly Bottle
Grade Reagent