missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trisodium N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetate Dihydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 288°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Formula Weight | 344.21 |
| Percent Purity | ≥98.0% (T) |
| Chemical Name or Material | Trisodium N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetate Dihydrate |
Safety and Handling
| EINECSNumber | (2)-1268 |
| RTECSNumber | MB9188000 |
| TSCA | Yes |