missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trisodium Hydrogen Ethylenediaminetetraacetate Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 358.19 |
| Percent Purity | ≥98.0% (T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Trisodium Hydrogen Ethylenediaminetetraacetate Hydrate [for Biochemical Research] |
Safety and Handling
| EINECSNumber | (2)-1265 |
| TSCA | Yes |