missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS, USP grade, 99.0 to 101.0%, MP Biomedicals™
Tris is excellent biochemical and biological buffer for all applications where ultra high purity is required
$88.90 - $329.73
Chemical Identifiers
| CAS | 77-86-1 |
|---|---|
| Molecular Formula | C4H11NO3 |
| Molecular Weight (g/mol) | 121.136 |
| InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
| Synonym | Tris-[hydroxymethyl]aminomethane, Trometamol |
| PubChem CID | 6503 |
| ChEBI | CHEBI:9754 |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
| SMILES | C(C(CO)(CO)N)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19560580
|
MP Biomedicals Inc
0219560580 |
100 g |
N/A
|
|
|||||
|
ICN19560590
|
MP Biomedicals Inc
0219560590 |
500 g |
N/A
|
|
|||||
|
ICN19560591
|
MP Biomedicals Inc
0219560591 |
1 kg |
N/A
|
|
|||||
Chemical Identifiers
| 77-86-1 | |
| 121.136 | |
| Tris-[hydroxymethyl]aminomethane, Trometamol | |
| CHEBI:9754 | |
| C(C(CO)(CO)N)O |
| C4H11NO3 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 6503 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol |
Specifications
| 77-86-1 | |
| Crystalline Powder | |
| 99.0 to 101.0% | |
| Trometamol, Tris-[hydroxymethyl]aminomethane | |
| C(C(CO)(CO)N)O | |
| 121.136 | |
| CHEBI:9754 | |
| USP |
| White | |
| 100 g | |
| C4H11NO3 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 6503 | |
| 121.14 | |
| TRIS |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30°C). Desiccate.