missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS, USP grade, 99.0 to 101.0%, MP Biomedicals™
Tris is excellent biochemical and biological buffer for all applications where ultra high purity is required
Supplier: MP Biomedicals Inc 0219560590
Specifications
| TRIS | |
| White | |
| 500 g | |
| C4H11NO3 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 6503 | |
| 121.14 |
| 77-86-1 | |
| Crystalline Powder | |
| 99.0 to 101.0% | |
| Tris-[hydroxymethyl]aminomethane, Trometamol | |
| C(C(CO)(CO)N)O | |
| 121.136 | |
| CHEBI:9754 | |
| USP |
Chemical Identifiers
| 77-86-1 | |
| 121.136 | |
| Tris-[hydroxymethyl]aminomethane, Trometamol | |
| CHEBI:9754 | |
| C(C(CO)(CO)N)O |
| C4H11NO3 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 6503 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30°C). Desiccate.