missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris, Ultra Pure grade, 99.95% min., MP Biomedicals™
$272.74 - $2027.32
Chemical Identifiers
| CAS | 77-86-1 |
|---|---|
| Molecular Formula | C4H11NO3 |
| Molecular Weight (g/mol) | 121.136 |
| MDL Number | MFCD00004679 |
| InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
| Synonym | 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, Tris base |
| PubChem CID | 6503 |
| ChEBI | CHEBI:9754 |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
| SMILES | C(C(CO)(CO)N)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10313301
|
MP Biomedicals Inc
0210313301 |
1 kg |
Each for $272.74
|
|
|||||
|
ICN10313305
|
MP Biomedicals Inc
0210313305 |
5 kg |
Each for $1,035.43
|
|
|||||
|
ICN10313310
|
MP Biomedicals Inc
0210313310 |
10 kg |
Each for $2,027.32
|
|
|||||
Chemical Identifiers
| 77-86-1 | |
| 121.136 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 6503 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol |
| C4H11NO3 | |
| MFCD00004679 | |
| 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, Tris base | |
| CHEBI:9754 | |
| C(C(CO)(CO)N)O |
Specifications
| 77-86-1 | |
| Crystalline Powder | |
| >99.9% | |
| MFCD00004679 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 6503 | |
| 121.1 | |
| Tris |
| White | |
| 1 kg | |
| C4H11NO3 | |
| 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, Tris base | |
| C(C(CO)(CO)N)O | |
| 121.136 | |
| CHEBI:9754 | |
| Ultrapure |
Safety and Handling
Recommended Storage : Store at room temperature, desiccate.