missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris, Ultra Pure grade, 99.95% min., MP Biomedicals™
Supplier: MP Biomedicals Inc 0210313305
Spécifications
| Tris | |
| White | |
| 5 kg | |
| C4H11NO3 | |
| 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, Tris base | |
| C(C(CO)(CO)N)O | |
| 121.136 | |
| CHEBI:9754 | |
| Ultrapure |
| 77-86-1 | |
| Crystalline Powder | |
| >99.9% | |
| MFCD00004679 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 6503 | |
| 121.1 |
Identifiants chimiques
| 77-86-1 | |
| 121.136 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 6503 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol |
| C4H11NO3 | |
| MFCD00004679 | |
| 2-Amino-2-(hydroxymethyl)-1, 3-propanediol, Tris base | |
| CHEBI:9754 | |
| C(C(CO)(CO)N)O |
Sécurité et manipulation
Recommended Storage : Store at room temperature, desiccate.