missing translation for 'onlineSavingsMsg'
Learn More

Tris (Tromethamine) (Granular/USP), Fisher Chemical™

Catalog No. T393500
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
T393500 500 g Poly Bottle
T393212 2.5 kg Poly Bottle
T39312 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T393500 Supplier Thermo Fisher Scientific Supplier No. FLT393 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Trishydroxymethylaminomethane, Tris Buffer, Tris, THAM, Trizma™, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Melting Point 168.5°C
Boiling Point 219°C
Color White
pH 10.4
Physical Form Powder or Solid
Quantity 500 g
Identification Pass Test (USP)
Loss on Drying 1.0% max. (USP)
Linear Formula H2NC(CH2OH)3
Packaging Poly Bottle
Formula Weight 121.14
Percent Purity 99.0 to 101.0% (USP)
Grade USP
Chemical Name or Material Tris (Tromethamine)
Show More Show Less