missing translation for 'onlineSavingsMsg'
Learn More

Tris(pentafluorophenyl)phosphine 95.0+%, TCI America™

Catalog No. T24845G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24845G 5 g
T24841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24845G Supplier TCI America Supplier No. T24845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1259-35-4
Molecular Formula C18F15P
Molecular Weight (g/mol) 532.15
MDL Number MFCD00079654
InChI Key FQLSDFNKTNBQLC-UHFFFAOYSA-N
Synonym tris pentafluorophenyl phosphine, phosphine, tris pentafluorophenyl, tris perfluorophenyl phosphine, tris pentafluorophenyl phosphane, tris 2,3,4,5,6-pentafluorophenyl phosphane, pubchem6422, acmc-1bogg, ksc491k1f, tris-pentafluorophenyl-phosphane
PubChem CID 102076
IUPAC Name tris(2,3,4,5,6-pentafluorophenyl)phosphane
SMILES FC1=C(F)C(F)=C(P(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 5 g
Formula Weight 532.15
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(pentafluorophenyl)phosphine