missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane hydrochloride, 99+%, extra pure

Catalog No. AC228031000
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228031000 100 g Plastic bottle
AC228032500 250 g Plastic bottle
AC228030010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC228031000 Supplier Thermo Scientific Chemicals Supplier No. 228031000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1185-53-1 | C4H11NO3·HCl | 157.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H11NO3·HCl
Molecular Weight (g/mol) 157.6
MDL Number MFCD00012590
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym tris hydrochloride, tris hydroxymethyl aminomethane hydrochloride, 2-amino-2-hydroxymethyl propane-1,3-diol hydrochloride, tromethamine hydrochloride, tris hcl, Tris-HCl, 2-amino-2-hydroxymethyl-1,3-propanediol hydrochloride, tris buffer hydrochloride, 1,3-propanediol, 2-amino-2-hydroxymethyl-, hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Melting Point 150.0°C to 153.0°C
Color White
pH 3.5 to 5 (1% soln. at 25°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula (HOCH2)3CNH2·HCl
Quantity 100 g
Solubility Information (1% in water) Clear colorless
Formula Weight 157.6
Percent Purity 99+%
Grade Extra Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tris(hydroxymethyl)aminomethane hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
EINECSNumber 214-684-5

RUO – Research Use Only