missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Tris, Hydrochloride, ULTROL™ Grade, Calbiochem™,

Catalog No. p-4953821
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
648313250GM 250 g
648313500GM 500 g
6483131KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 648313250GM Supplier MilliporeSigma™ Supplier No. 648313250GM
Only null left

Useful in the pH of 3.5 to 5.0

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym tris(Hydroxymethyl)aminomethane, HCl
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Melting Point 149°C
Quantity 250 g
Solubility Information Soluble in water (800g/L at 20°C)
Formula Weight 157.6
Chemical Name or Material Tris, Hydrochloride
Grade by titration
Recommended Storage 15°C to 30°C (Do not freeze).