missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Tris, Hydrochloride, ULTROL™ Grade, Calbiochem™,
Specifications
Specifications
| Melting Point | 149°C |
| For Use With (Application) | Laboratory |
| pH | 4.2 |
| Physical Form | Solid |
| Quantity | 500 g |
| Solution Type | Buffer |
| Solubility Information | Soluble in water (800g/L at 20°C) |
| Formula Weight | 157.6 |
| Percent Purity | ≥99% (titration) |
| Grade | by titration |
| Show More |
Safety and Handling
| Recommended Storage | 15°C to 30°C (Do not freeze). |