missing translation for 'onlineSavingsMsg'
Learn More

Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents

Catalog No. bp1531
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1531 1 kg Poly Bottle
BP153500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP1531 Supplier Fisher BioReagents Supplier No. BP153 1
Only null left

For electrophoresis, molecular biology, and cell culture applications | CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

For electrophoresis, molecular biology, and cell culture applications

Tris Hydrochloride is a buffering medium for electrophoresis, molecular biology, and cell culture applications. It may be combined with Tris Base to simplify Tris buffer preparation.

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym Tris(hydroxymethyl)aminomethane Hydrochloride
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Absorbance 0.05 max. (1M solution) at 280nm
Melting Point 150°C
Color White
pH 3.5 to 5.0
Physical Form Solid
Quantity 1 kg
DNase DNase free
Formulation Tris (hydroxymethyl) Aminomethane, Hydrochloride
Assay Percent Range ≥99 %
Identification Pass Test
Linear Formula H2NC(CH2OH)3·HCl
Packaging Poly Bottle
Protease Protease free
Formula Weight 157.6
Percent Purity ≥99%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Tris Hydrochloride
Show More Show Less
Recommended Storage Keep in a dry, cool and well-ventilated place. Keep container tightly closed.