missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ TRIS Hydrochloride, OmniPur™, Calbiochem™,

Catalog No. 9310500GM
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9310500GM 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 9310500GM Supplier MilliporeSigma™ Supplier No. 9310500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity ULTROL™ grade buffer for sensitive biological and biochemical applications.

Recommended Storage

Ok to freeze

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym Trizma, Tris(hydroxymethyl)aminomethane
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Color White
For Use With (Application) Molecular Biology
pH 3.5 to 5.0
Physical Form Crystalline Powder to Needles
Quantity 500 g
Solution Type Buffer
Packaging Fiber Carton
Formula Weight 157.6
Percent Purity ≥99.0%
Grade Molecular Biology
Chemical Name or Material TRIS Hydrochloride
Show More Show Less
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use