missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Tris Hydrochloride, OmniPur™, Calbiochem™,
Specifications
Specifications
| Physical Form | Crystalline |
| Quantity | 5 kg |
| DNase | None Detected |
| Protease | None Detected |
| Formula Weight | 157.6 |
| Chemical Name or Material | Tris Hydrochloride |
Safety and Handling
| Recommended Storage | Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. |