missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Tris, Hydrochloride, Molecular Biology Grade, Calbiochem™,

Catalog No. p-4953820
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
648317100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 648317100GM Supplier MilliporeSigma™ Supplier No. 648317100GM
Only null left

Useful in the pH range of 3.5 to 5.0

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
Synonym tris(Hydroxymethyl)aminomethane, HCl
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Melting Point 149°C
Quantity 100 g
DNase None Detected
Assay Percent Range ≥99.0%
Protease None Detected
Solubility Information Easily soluble in cold water
Formula Weight 157.6
Grade by titration
Chemical Name or Material Tris, Hydrochloride
Recommended Storage 15°C to 30°C (Do not freeze).