missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Tris, Hydrochloride, Molecular Biology Grade, Calbiochem™,
Specifications
Specifications
| Melting Point | 149°C |
| For Use With (Application) | Molecular Biology |
| pH | 7.0 to 9.0 |
| Physical Form | Crystalline |
| Quantity | 100 g |
| Solution Type | Buffer |
| DNase | None Detected |
| Assay Percent Range | ≥99.0% |
| Protease | None Detected |
| Solubility Information | Easily soluble in cold water |
| Show More |
Safety and Handling
| Recommended Storage | 15°C to 30°C (Do not freeze). |