missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Hydrochloride, >99%, Reagent grade, MP Biomedicals™
$538.75 - $1900.99
Chemical Identifiers
| CAS | 1185-53-1 |
|---|---|
| Molecular Formula | C4H12ClNO3 |
| Molecular Weight (g/mol) | 157.594 |
| MDL Number | MFCD00012590 |
| InChI Key | QKNYBSVHEMOAJP-UHFFFAOYSA-N |
| Synonym | Trometamol, Tromethamine |
| PubChem CID | 93573 |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
| SMILES | C(C(CO)(CO)N)O.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10313001
|
MP Biomedicals Inc
0210313001 |
1 kg |
Each for $538.75
|
|
|||||
|
ICN10313005
|
MP Biomedicals Inc
0210313005 |
5 kg |
Each for $1,900.99
|
|
|||||
Description
Cell Biology and AnalysisChemical Identifiers
| 1185-53-1 | |
| 157.594 | |
| QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
| 93573 | |
| C(C(CO)(CO)N)O.Cl |
| C4H12ClNO3 | |
| MFCD00012590 | |
| Trometamol, Tromethamine | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
Specifications
| 1185-53-1 | |
| Crystalline Powder | |
| >99% | |
| MFCD00012590 | |
| QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
| 93573 | |
| Reagent |
| White | |
| 1 kg | |
| C4H12ClNO3 | |
| Trometamol, Tromethamine | |
| C(C(CO)(CO)N)O.Cl | |
| 157.594 | |
| 157.6 | |
| Tris Hydrochloride |
Safety and Handling
Recommended Storage : Store at room temperature (15° to 30°C).