missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Hydrochloride, >99%, Reagent grade, MP Biomedicals™
Description
Cell Biology and Analysis
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 1 kg |
| Assay Percent Range | >99% |
| Formula Weight | 157.6 |
| Grade | Reagent |
| Chemical Name or Material | Tris Hydrochloride |
Safety and Handling
| Recommended Storage | Store at room temperature (15° to 30°C). |