missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris-HCl, Molecular Grade, Promega™
Used in combination with Tris base for preparation of Tris-HCl buffers.
$96.19 - $878.84
Chemical Identifiers
| CAS | 1185-53-1 |
|---|---|
| Molecular Formula | C4H12ClNO3 |
| Molecular Weight (g/mol) | 157.594 |
| InChI Key | QKNYBSVHEMOAJP-UHFFFAOYSA-N |
| PubChem CID | 93573 |
| IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
| SMILES | C(C(CO)(CO)N)O.Cl |
Description
- Quality tested and certified free of DNase, RNase and protease activity
Chemical Identifiers
| 1185-53-1 | |
| 157.594 | |
| 93573 | |
| C(C(CO)(CO)N)O.Cl |
| C4H12ClNO3 | |
| QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride |
Specifications
| 1185-53-1 | |
| C4H12ClNO3 | |
| C(C(CO)(CO)N)O.Cl | |
| 157.594 | |
| 157.56 | |
| Tris-HCl |
| 100 g | |
| QKNYBSVHEMOAJP-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride | |
| 93573 | |
| Molecular Biology |
Safety and Handling
Recommended Storage : 15 to 30°C