missing translation for 'onlineSavingsMsg'
Learn More

Tris-HCl, 1M Solution, pH 8.0, Molecular Biology Grade, Ultrapure

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Supplier:  Thermo Fisher Scientific J22638AP

 View more versions of this product

Catalog No. AAJ22638AP


Only null left

Description

Description

Specifications

Specifications

Tris-HCl
1 M
Solution
C4H12ClNO3
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
93573
Molecular Biology
1185-53-1
8
500 mL
Nalgene bottle
C(C(CO)(CO)N)O.Cl
157.594
157.6

Chemical Identifiers

1185-53-1
157.594
93573
C(C(CO)(CO)N)O.Cl
C4H12ClNO3
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only