missing translation for 'onlineSavingsMsg'
Learn More

Tris-HCl, 1M Solution, pH 8.0, Molecular Biology Grade, Ultrapure

Catalog No. AAJ22638AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ22638AE 100 mL Nalgene bottle
AAJ22638AP 500 mL Nalgene bottle
AAJ22638K2 1 L Nalgene bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ22638AE Supplier Thermo Fisher Scientific Supplier No. J22638AE

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Concentration 1 M
pH 8
Physical Form Solution
Quantity 100 mL
Packaging Nalgene bottle
Formula Weight 157.6
Grade Molecular Biology
Chemical Name or Material Tris-HCl
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only