missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris-HCl, 0.5M buffer soln., pH 8.0, low endotoxin

Catalog No. p-7057903
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67510AE 100 mL
AAJ67510AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67510AE Supplier Thermo Scientific Chemicals Supplier No. J67510AE

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00012590
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Color Colorless
Concentration 0.5 M
pH 8.0
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material TRIS-HCl, low endotoxin
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only