missing translation for 'onlineSavingsMsg'
Learn More

Tris(ethylenediamine)cobalt chloride trihydrate, 97%, Thermo Scientific™

Catalog No. AC458960050
Change view
Click to view available options
Quantity:
5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458960050 5g
AC458960250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458960050 Supplier Thermo Scientific Chemicals Supplier No. 458960050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14883-80-8
Molecular Formula C6H24Cl3CoN6·3H2O
Molecular Weight (g/mol) 399.64
InChI Key WBMVTRUQSMZIKW-UHFFFAOYSA-K
Synonym tris ethylenediamine cobalt iii chloride trihydrate, cobalt 3+ tris ethylenediamine trihydrate trichloride
PubChem CID 73995044
IUPAC Name cobalt(3+);ethane-1,2-diamine;trichloride;trihydrate
SMILES C(CN)N.C(CN)N.C(CN)N.O.O.O.[Cl-].[Cl-].[Cl-].[Co+3]

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Quantity 5g
Solubility Information Solubility in water: soluble.
Formula Weight 399.64
Percent Purity 97%
Chemical Name or Material Tris(ethylenediamine)cobalt chloride trihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse

RUO – Research Use Only