missing translation for 'onlineSavingsMsg'
Learn More

Tris(dibenzoylmethanato) Iron 97.0+%, TCI America™

Catalog No. T16865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16865G 5 g
T168625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16865G Supplier TCI America Supplier No. T16865G

Chemical Identifiers

CAS 14405-49-3
Molecular Formula C45H33FeO6
Molecular Weight (g/mol) 725.60
MDL Number MFCD00192557,MFCD14636458
InChI Key JWPHPBLJVLHKOZ-UHFFFAOYSA-K
Synonym tris dibenzoylmethanato iron, iron iii diphenylpropanedionate
PubChem CID 57369570
IUPAC Name iron(3+) tris(3-oxo-1,3-diphenylprop-1-en-1-olate)
SMILES [Fe+3].[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1.[O-]C(=CC(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 725.60
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(dibenzoylmethanato) Iron