missing translation for 'onlineSavingsMsg'
Learn More

Tris(8-quinolinolato)aluminum (purified by sublimation) 98.0+%, TCI America™

Catalog No. T22385G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T22385G Supplier TCI America Supplier No. T22385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2085-33-8
Molecular Formula C27H18AlN3O3
MDL Number MFCD00191693
Synonym tri-8-quinolinolatoaluminum, aluminum 8-hydroxyquinolinate, tri quinolin-8-yloxy alumane, aluminum tris quinolin-8-olate, tris 8-hydroxyquinolinato aluminum, aluminum, tris 8-quinolinato-n1,o8, aluminum oxinate, aluminum oxinate,, tris 8-quinolyloxy alumane, tris 8-quinolinyloxy alumane

Specifications

Melting Point 418°C
Quantity 5 g
Formula Weight 459.44
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(8-quinolinolato)aluminum (purified by sublimation)
RTECSNumber BD2231000
TSCA No