missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-fluorophenyl)phosphine 98.0+%, TCI America™

Catalog No. T29005G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T29005G Supplier TCI America Supplier No. T29005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18437-78-0
Molecular Formula C18H12F3P
Molecular Weight (g/mol) 316.26
MDL Number MFCD00013553
InChI Key GEPJPYNDFSOARB-UHFFFAOYSA-N
Synonym tris 4-fluorophenyl phosphine, phosphine, tris 4-fluorophenyl, tris p-fluorophenyl phosphine, tri 4-fluorophenyl phosphine, tris 4-fluorophenyl phosphane, phosphine, tris p-fluorophenyl, zlchem 700, pubchem6418, acmc-1c3oi
PubChem CID 140387
IUPAC Name tris(4-fluorophenyl)phosphane
SMILES FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1

Specifications

Melting Point 81°C
Color White
Boiling Point 160°C
Quantity 5 g
Formula Weight 316.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-fluorophenyl)phosphine